Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ing5
Entrez Gene ID
363292
Gene Name
inhibitor of growth family, member 5
Gene Aliases
-
Chromosome Location
Chr.9: 100932932 - 100950233 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108810.1 NP_001102280.1 4 - 5 527 63

View Full Product Details