The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108810.1 | NP_001102280.1 | 7 - 8 | 813 | 126 |
GenBank mRNA | BC167048.1 | - | 7 - 8 | 683 | 126 |