The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003750733.2 | XP_003750781.1 | 2 - 3 | 940 | 72 |
GenBank mRNA | FQ216933.1 | - | 2 - 3 | 908 | 72 |
FQ216966.1 | - | 2 - 3 | 940 | 72 |