Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet1
Entrez Gene ID
309902
Gene Name
tet methylcytosine dioxygenase 1
Gene Aliases
Cxxc6
Chromosome Location
Chr.20: 27359125 - 27438039 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256398.2 XP_006256460.1 11 - 12 5359 77

View Full Product Details