Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-Ba
Entrez Gene ID
309621
Gene Name
RT1 class II, locus Ba
Gene Aliases
BA1
Chromosome Location
Chr.20: 4066133 - 4070726 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008831.3 NP_001008831.1 3 - 4 681 82
GenBank mRNA AY626197.1 - 3 - 4 619 82
BC086448.1 - 3 - 4 631 82
FQ225318.1 - 3 - 4 673 82
FQ225365.1 - 3 - 4 673 82
FQ225680.1 - 3 - 4 663 82
FQ225905.1 - 3 - 4 673 82
FQ226059.1 - 3 - 4 673 82
FQ226209.1 - 3 - 4 82
FQ226311.1 - 3 - 4 674 82
FQ227679.1 - 3 - 4 673 82
FQ227967.1 - 3 - 4 634 82
FQ228231.1 - 3 - 4 673 82
FQ229986.1 - 3 - 4 673 82
FQ231104.1 - 3 - 4 652 82
FQ231505.1 - 3 - 4 634 82
FQ231715.1 - 3 - 4 673 82
FQ231787.1 - 3 - 4 673 82
FQ231789.1 - 3 - 4 673 82
FQ232205.1 - 3 - 4 676 82
FQ232339.1 - 3 - 4 673 82
FQ232777.1 - 3 - 4 673 82
FQ232809.1 - 3 - 4 673 82
FQ233515.1 - 3 - 4 670 82
FQ234579.1 - 3 - 4 663 82
FQ234725.1 - 3 - 4 673 82
KC222945.1 - 3 - 4 619 82

View Full Product Details