Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tulp1
Entrez Gene ID
309900
Gene Name
tubby like protein 1
Gene Aliases
-
Chromosome Location
Chr.20: 7931673 - 7943604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107642.1 NP_001101112.1 10 - 11 1051 77
XM_006256217.2 XP_006256279.1 12 - 13 1311 77
XM_006256218.2 XP_006256280.1 11 - 12 1305 77
XM_006256219.2 XP_006256281.1 12 - 13 1311 77

View Full Product Details