Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtch1
Entrez Gene ID
294313
Gene Name
mitochondrial carrier 1
Gene Aliases
-
Chromosome Location
Chr.20: 6937341 - 6961165 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100833.1 NP_001094303.1 6 - 7 709 104
XM_006256184.1 XP_006256246.1 6 - 7 731 104
GenBank mRNA BC158661.1 - 6 - 7 709 104

View Full Product Details