Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ring1
Entrez Gene ID
309626
Gene Name
ring finger protein 1
Gene Aliases
Ring1A
Chromosome Location
Chr.20: 3812287 - 3815834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212549.2 NP_997714.2 4 - 5 658 148
GenBank mRNA AB568263.1 - 3 - 4 442 148
BC098635.1 - 4 - 5 602 148

View Full Product Details