The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212549.2 | NP_997714.2 | 5 - 6 | 1035 | 73 |
GenBank mRNA | AB568263.1 | - | 4 - 5 | 819 | 73 |
BC098635.1 | - | 5 - 6 | 979 | 73 |