Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upb1
Entrez Gene ID
116593
Gene Name
ureidopropionase, beta
Gene Aliases
Bup1
Chromosome Location
Chr.20: 14167383 - 14193724 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053845.1 NP_446297.1 7 - 8 900 87
GenBank mRNA BC078767.1 - 7 - 8 890 87
FQ209442.1 - 7 - 8 878 87
M97662.1 - 7 - 8 900 87

View Full Product Details