The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013906.1 | NP_001013928.1 | 1 - 2 | 656 | 135 |
GenBank mRNA | BC087673.1 | - | 1 - 2 | 656 | 135 |