Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfdn6
Entrez Gene ID
309629
Gene Name
prefoldin subunit 6
Gene Aliases
Ke2
Chromosome Location
Chr.20: 5455974 - 5457448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164718.1 NP_001158190.1 2 - 3 191 72
NM_212506.2 NP_997671.1 1 - 2 300 72

View Full Product Details