Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tesb
Entrez Gene ID
414826
Gene Name
testis specific basic protein
Gene Aliases
-
Chromosome Location
Chr.20: 4178662 - 4235947 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113183.1 NP_001106654.1 1 - 2 151 115
XM_008772756.1 XP_008770978.1 5 - 6 469 115
GenBank mRNA BC083628.1 - 1 - 2 150 115
BC097390.1 - 1 - 2 151 115

View Full Product Details