The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106759.2 | NP_001100229.1 | 11 - 12 | 2274 | 82 |
XM_006240572.2 | XP_006240634.1 | 11 - 12 | 2274 | 82 | |
XM_006240573.2 | XP_006240635.1 | 11 - 12 | 2232 | 82 | |
XM_006240574.2 | XP_006240636.1 | 11 - 12 | 4652 | 82 | |
XM_006240575.2 | XP_006240637.1 | 11 - 12 | 2378 | 82 | |
XM_006240576.2 | XP_006240638.1 | 11 - 12 | 2041 | 82 | |
GenBank mRNA | BC166426.1 | - | 11 - 12 | 2274 | 82 |
FQ230530.1 | - | 11 - 12 | 2293 | 82 |