Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exoc3l4
Entrez Gene ID
299338
Gene Name
exocyst complex component 3-like 4
Gene Aliases
RGD1307749
Chromosome Location
Chr.6: 135866138 - 135880724 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106759.2 NP_001100229.1 9 - 10 1995 91
XM_006240572.2 XP_006240634.1 9 - 10 1995 91
XM_006240573.2 XP_006240635.1 9 - 10 1953 91
XM_006240574.2 XP_006240636.1 9 - 10 4373 91
XM_006240575.2 XP_006240637.1 9 - 10 2099 91
XM_006240576.2 XP_006240638.1 9 - 10 1762 91
GenBank mRNA BC166426.1 - 9 - 10 1995 91
FQ230530.1 - 9 - 10 2014 91

View Full Product Details