The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100894.1 | NP_001094364.1 | 1 - 2 | 383 | 95 |
XM_006240583.2 | XP_006240645.1 | 1 - 2 | 319 | 95 | |
XM_008764933.1 | XP_008763155.1 | 1 - 2 | 321 | 95 | |
XM_008764934.1 | XP_008763156.1 | 1 - 2 | 321 | 95 | |
XM_008764935.1 | XP_008763157.1 | 1 - 2 | 323 | 95 |