Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9a7
Entrez Gene ID
317170
Gene Name
solute carrier family 9, subfamily A (NHE7, cation proton antiporter 7), member 7
Gene Aliases
Nhe7
Chromosome Location
Chr.X: 2434914 - 2615314 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108242.1 NP_001101712.1 14 - 15 1864 63
XM_006256636.2 XP_006256698.1 14 - 15 2245 63
XM_006256637.2 XP_006256699.1 13 - 14 2185 63

View Full Product Details