Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Araf
Entrez Gene ID
64363
Gene Name
A-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
Araf1
Chromosome Location
Chr.X: 1369591 - 1391615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033663.2 NP_001028835.1 1 - 2 51 124
NM_022532.2 NP_071977.1 1 - 2 51 124
XM_008773036.1 XP_008771258.1 1 - 2 793 124
GenBank mRNA CO572305.1 - 1 - 2 51 124

View Full Product Details