Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc29
Entrez Gene ID
502201
Gene Name
leucine rich repeat containing 29
Gene Aliases
-
Chromosome Location
Chr.19: 37266782 - 37282076 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195616.1 NP_001182545.1 4 - 5 1293 76
XM_006255531.2 XP_006255593.1 4 - 5 1528 76
XM_006255532.2 XP_006255594.1 4 - 5 1436 76
XM_006255533.2 XP_006255595.1 4 - 5 1372 76
GenBank mRNA BC166813.1 - 4 - 5 1293 76

View Full Product Details