Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsf3
Entrez Gene ID
498962
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
Chromosome Location
Chr.19: 55594741 - 55635312 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_574249.6 XP_574249.4 1 - 2 188 64

View Full Product Details