Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk10
Entrez Gene ID
361434
Gene Name
cyclin-dependent kinase 10
Gene Aliases
-
Chromosome Location
Chr.19: 56024903 - 56032610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025722.2 NP_001020893.2 4 - 5 874 103
NM_001109936.1 NP_001103406.1 5 - 6 423 103
NM_001109937.1 NP_001103407.1 5 - 6 384 103
GenBank mRNA BC098804.1 - 4 - 5 874 103
DN932869.1 - 5 - 6 419 103
DN935585.1 - 5 - 6 443 103
DY320517.1 - 5 - 6 424 103

View Full Product Details