Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk10
Entrez Gene ID
361434
Gene Name
cyclin-dependent kinase 10
Gene Aliases
-
Chromosome Location
Chr.19: 56024903 - 56032610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025722.2 NP_001020893.2 10 - 11 1397 72
NM_001109936.1 NP_001103406.1 11 - 12 946 72
NM_001109937.1 NP_001103407.1 11 - 12 907 72
GenBank mRNA BC098804.1 - 10 - 11 1397 72

View Full Product Details