Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anapc10
Entrez Gene ID
361389
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
-
Chromosome Location
Chr.19: 31814686 - 31867928 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108445.1 NP_001101915.1 4 - 5 498 99
XM_006255406.2 XP_006255468.1 4 - 5 500 99
GenBank mRNA BC168746.1 - 4 - 5 420 99

View Full Product Details