Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adad2
Entrez Gene ID
691275
Gene Name
adenosine deaminase domain containing 2
Gene Aliases
RGD1560949
Chromosome Location
Chr.19: 52258908 - 52263031 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772652.1 XP_008770874.1 1 - 2 391 63

View Full Product Details