Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znrf1
Entrez Gene ID
690769
Gene Name
zinc and ring finger 1, E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.19: 43750205 - 43834962 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751882.3 XP_003751930.1 4 - 5 941 90
XM_006255651.2 XP_006255713.1 5 - 6 1043 90

View Full Product Details