Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gab1
Entrez Gene ID
361388
Gene Name
GRB2-associated binding protein 1
Gene Aliases
-
Chromosome Location
Chr.19: 30794290 - 30903819 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108444.1 NP_001101914.1 7 - 8 1964 113
GenBank mRNA GQ140625.1 - 8 - 9 1764 113

View Full Product Details