Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxf1
Entrez Gene ID
687536
Gene Name
forkhead box F1
Gene Aliases
-
Chromosome Location
Chr.19: 53012306 - 53016086 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006255724.2 XP_006255786.1 1 - 2 1005 53

View Full Product Details