The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107160.1 | NP_001100630.1 | 3 - 4 | 569 | 109 |
GenBank mRNA | BC158705.1 | - | 3 - 4 | 495 | 109 |