Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osgin1
Entrez Gene ID
171493
Gene Name
oxidative stress induced growth inhibitor 1
Gene Aliases
Okl38
Chromosome Location
Chr.19: 52056816 - 52085491 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138504.2 NP_612513.1 7 - 8 1026 86
XM_006255706.2 XP_006255768.1 10 - 11 1085 86
XM_006255707.2 XP_006255769.1 6 - 7 1121 86
XM_008772594.1 XP_008770816.1 5 - 6 947 86
GenBank mRNA AF549441.1 - 7 - 8 1026 86
AF549442.1 - 6 - 7 927 86
AY081218.1 - 5 - 6 675 86
BC091111.1 - 7 - 8 1022 86

View Full Product Details