Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scoc
Entrez Gene ID
364981
Gene Name
short coiled-coil protein
Gene Aliases
-
Chromosome Location
Chr.19: 24584435 - 24614170 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013235.1 NP_001013253.1 2 - 3 159 93
XM_006255293.2 XP_006255355.1 2 - 3 188 93
GenBank mRNA BC076390.1 - 2 - 3 159 93
BC086417.1 - 2 - 3 159 93

View Full Product Details