The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025766.1 | NP_001020937.1 | 4 - 5 | 774 | 67 |
XM_006255599.2 | XP_006255661.1 | 5 - 6 | 1140 | 67 | |
XM_006255600.2 | XP_006255662.1 | 5 - 6 | 1137 | 67 | |
XM_008772527.1 | XP_008770749.1 | 5 - 6 | 845 | 67 | |
GenBank mRNA | BC076391.1 | - | 5 - 6 | 903 | 67 |
BC098808.1 | - | 4 - 5 | 774 | 67 |