Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pabpn1l
Entrez Gene ID
307920
Gene Name
poly(A)binding protein nuclear 1-like
Gene Aliases
Epabp2, Pabpnl1, RGD1562761
Chromosome Location
Chr.19: 55430808 - 55434252 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113779.1 NP_001107251.1 4 - 5 582 80
GenBank mRNA BC158789.1 - 4 - 5 582 80

View Full Product Details