Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsd17b2
Entrez Gene ID
79243
Gene Name
hydroxysteroid (17-beta) dehydrogenase 2
Gene Aliases
-
Chromosome Location
Chr.19: 50246404 - 50317892 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC088134.1 - 3 - 4 708 97

View Full Product Details