Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbfa2t3
Entrez Gene ID
361431
Gene Name
core-binding factor, runt domain, alpha subunit 2; translocated to, 3
Gene Aliases
-
Chromosome Location
Chr.19: 55438409 - 55512131 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108453.1 NP_001101923.1 6 - 7 904 80
XM_006255770.2 XP_006255832.1 5 - 6 1798 80
XM_006255772.2 XP_006255834.1 6 - 7 1059 80
XM_008772629.1 XP_008770851.1 5 - 6 2769 80
XM_008772630.1 XP_008770852.1 4 - 5 2692 80
XM_008772631.1 XP_008770853.1 5 - 6 1254 80

View Full Product Details