Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nepn
Entrez Gene ID
309775
Gene Name
nephrocan
Gene Aliases
RGD1307755
Chromosome Location
Chr.20: 33527932 - 33543860 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256491.2 XP_006256553.1 1 - 2 395 89

View Full Product Details