Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vgll2
Entrez Gene ID
309772
Gene Name
vestigial-like family member 2
Gene Aliases
-
Chromosome Location
Chr.20: 33077106 - 33082962 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107630.1 NP_001101100.1 2 - 3 582 103
XM_008772885.1 XP_008771107.1 2 - 3 1195 103

View Full Product Details