Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sept10
Entrez Gene ID
309891
Gene Name
septin 10
Gene Aliases
-
Chromosome Location
Chr.20: 28722446 - 28814841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014033.1 NP_001014055.1 8 - 9 1208 162
XM_006256415.2 XP_006256477.1 9 - 10 1433 162
XM_006256417.2 XP_006256479.1 8 - 9 1426 162
XM_006256418.2 XP_006256480.1 9 - 10 1432 162
XM_008772886.1 XP_008771108.1 8 - 9 1711 162
GenBank mRNA BC087157.1 - 8 - 9 1208 162

View Full Product Details