The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024743.1 | NP_001019914.1 | 9 - 10 | 1662 | 105 |
XM_006251538.1 | XP_006251600.1 | 9 - 10 | 1614 | 105 | |
GenBank mRNA | BC084711.1 | - | 4 - 5 | 769 | 105 |
BC097369.1 | - | 9 - 10 | 1662 | 105 | |
FQ214781.1 | - | 4 - 5 | 844 | 105 |