The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107345.1 | NP_001100815.1 | 6 - 7 | 1291 | 127 |
XM_008771570.1 | XP_008769792.1 | 6 - 7 | 1028 | 127 |