Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d7
Entrez Gene ID
361227
Gene Name
TBC1 domain family, member 7
Gene Aliases
-
Chromosome Location
Chr.17: 23774793 - 23792396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108411.1 NP_001101881.1 3 - 4 400 67
XM_006253830.2 XP_006253892.1 3 - 4 421 67
XM_006253832.2 XP_006253894.1 2 - 3 378 67

View Full Product Details