Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mylip
Entrez Gene ID
306825
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
MIR
Chromosome Location
Chr.17: 19682040 - 19703681 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107344.2 NP_001100814.2 3 - 4 683 99

View Full Product Details