The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108576.1 | NP_001102046.1 | 2 - 3 | 228 | 154 |
XM_006233832.2 | XP_006233894.1 | 1 - 2 | 93 | 154 |