Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx3
Entrez Gene ID
170671
Gene Name
LIM homeobox 3
Gene Aliases
-
Chromosome Location
Chr.3: 3653861 - 3662522 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001059910.4 XP_001059910.2 1 - 2 189 97

View Full Product Details