Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcn8
Entrez Gene ID
366008
Gene Name
lipocalin 8
Gene Aliases
RGD1306747
Chromosome Location
Chr.3: 2871890 - 2875682 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128183.1 NP_001121655.1 1 - 2 118 75
XM_006233632.2 XP_006233694.2 1 - 2 259 75
GenBank mRNA DQ537491.1 - 1 - 2 104 75

View Full Product Details