Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspa5
Entrez Gene ID
25617
Gene Name
heat shock protein 5
Gene Aliases
BIP, GRP78
Chromosome Location
Chr.3: 13838304 - 13842763 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013083.2 NP_037215.1 1 - 2 76 67
GenBank mRNA BC062017.1 - 1 - 2 36 67
CB578015.1 - 1 - 2 76 67

View Full Product Details