Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
296623
Gene Name
family with sequence similarity 73, member B
Gene Aliases
RGD1310887
Chromosome Location
Chr.3: 8928495 - 8951189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106566.1 NP_001100036.1 9 - 10 1198 96
XM_006233781.1 XP_006233843.1 9 - 10 1247 96
XM_006233782.2 XP_006233844.1 9 - 10 1263 96
GenBank mRNA FQ212119.1 - 9 - 10 1166 96

View Full Product Details