Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stom
Entrez Gene ID
296655
Gene Name
stomatin
Gene Aliases
-
Chromosome Location
Chr.3: 14515576 - 14538199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011965.1 NP_001011965.1 2 - 3 194 103
GenBank mRNA BC083895.1 - 2 - 3 194 103

View Full Product Details