The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107825.1 | NP_001101295.1 | 6 - 7 | 1560 | 67 |
XM_006233800.2 | XP_006233862.1 | 8 - 9 | 1773 | 67 | |
XM_006233802.2 | XP_006233864.1 | 8 - 9 | 2082 | 67 | |
XM_006233804.2 | XP_006233866.1 | 5 - 6 | 1458 | 67 |