The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033884.1 | NP_001029056.1 | 4 - 5 | 944 | 85 |
GenBank mRNA | BC161820.1 | - | 4 - 5 | 909 | 85 |
CB611468.1 | - | 3 - 4 | 429 | 85 |