Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mms22l
Entrez Gene ID
313108
Gene Name
MMS22-like, DNA repair protein
Gene Aliases
RGD1304693
Chromosome Location
Chr.5: 38844494 - 38960989 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135780.1 NP_001129252.1 16 - 17 2924 80
XM_006237954.2 XP_006238016.1 16 - 17 2976 80

View Full Product Details